Learn More
LanthaScreen™ Eu-anti-His Tag Antibody Kit
LanthaScreen™ Eu-anti-His Tag Antibody Kit
$1628.00 - $19250.00
Specifications
| Antigen | His |
|---|---|
| Concentration | 0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml |
| Molecular Weight of Antigen | 150kDa |
| Packaging | 1 Kit(s) |
| Primary or Secondary | Primary |
Description
LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.Specifications
| His | |
| 150kDa | |
| Primary | |
| Liquid |
| 0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
| 1 Kit(s) | |
| Europium-labeled 6×His antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
| RUO |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.