Learn More
Invitrogen™ LanthaScreen™ Tb-anti-GST Antibody
LanthaScreen™ terbium-labeled anti-GST Goat antibodies are used to indirectly label proteins tagged with GST.
Supplier:  Invitrogen™ PV3551
Encompass_Preferred
 View more versions of this product
Catalog No. PV3551
Only null left
Edge
Add to Cart
Request a Quote
Description
Description
LanthaScreen™ terbium-labeled anti-GST Goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tb (Terbium)/Lanthanide Chelates
Terbium-labeled anti-GST antibody in HEPES buffered saline. Store at -20°C.
Dry Ice
LanthaScreen™
1 mg
340/490, 520 nm
End Point
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only. Not for use in diagnostic procedures.