Learn More
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
LanthaScreen™ Tb-anti-pSTAT1 [pTyr701] Antibody Kit
$5220.00 - $30280.00
Specifications
| Antigen | Tyrosine kinases |
|---|---|
| Packaging | 1 Kit(s) |
| Primary or Secondary | Primary |
| Content And Storage | Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C. |
| Regulatory Status | RUO |
Description
A part of the JAK/STAT Pathway, STAT1 (Signal Transducer and Activator of Transcription 1) is phosphorylated at Tyr701 by the Janus family of tyrosine kinases.For use in combination with the LanthaScreen™ STAT1 U206 cell line (K1469) and the GFP-STAT1 physiological protein substrate (PV5211) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Tyrosine kinases | |
| Primary | |
| RUO |
| 1 Kit(s) | |
| Terbium-labeled anti-STAT1 [pTyr701] antibody in HEPES buffered saline. Store at -20°C. |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.