Learn More
Invitrogen™ LanthaScreen™ Tb-peIF2a (pSer52) Antibody
The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis.
Supplier: Invitrogen™ PV4816

Description
The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis. In response to a cellular stress, phosphorylation of eIF2alpha at Ser52 occurs by the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2) to prohibit the initiation of protein synthesis.
For use in combination with the GFP-EIF2alpha physiological protein substrate (Cat. No. PV4809) to evaluate inhibitors in medium to high throughput screening applications.

Specifications
Tb (Terbium)/Lanthanide Chelates | |
Terbium-labeled peIF2a (pThr46) antibody in HEPES buffered saline. Store at -20°C. | |
Human | |
End Point | |
Ser/Thr Kinases |
1 mg | |
340/490, 520 nm | |
Dry Ice | |
Hu | |
LanthaScreen™ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.