Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LanthaScreen™ Tb-peIF2a (pSer52) Antibody Kit

The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis.

Supplier:  Invitrogen™ PV4810

Encompass_Preferred

 View more versions of this product

Catalog No. PV4810

  • $4,697.65 / Each of 1

    Save 6%

    Reg : $4,980.00

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart

Description

Description

The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis. In response to a cellular stress, phosphorylation of eIF2alpha at Ser52 occurs by the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2) to prohibit the initiation of protein synthesis.

For use in combination with the GFP-EIF2alpha physiological protein substrate (Cat. No. PV4809).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Serine/threonine kinase
Tb (Terbium)/GFP (EmGFP)
25 μg
Kinase Activity
9833
Terbium-labeled anti-eIF2alpha [pSer52] antibody in HEPES buffered saline. Store at -20°C.
Biochemical terbium phospho-specific activity
340/490, 520 nm
Dry Ice
Hu
Serine/Threonine Kinases
EIF2AK2, EIF2AK3
Kinase Assay, Enzyme and Protein Activity Assays, Protein Assay
HPK38, KIAA017
RUO
Primary
1 Each
150,000 Da
384-well Plate
Human
End Point
EIF2AK1, EIF2AK2, EIF2AK3
TR-FRET
LanthaScreen™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.