Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LanthaScreen™ Tb-PY20 Antibody Kit

LanthaScreen™ Tb-PY20 Antibody Kit

$1120.00

Specifications

Molecular Weight of Antigen 150kDa
Packaging 1 Kit(s)
Primary or Secondary Primary
Storage Requirements –20°C
Applications Cell Analysis, Cell Viability, Drug Discovery and Development, Kinase Antibodies and Immunoassays, Pharma and Biopharma, Proliferation and Function, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Validation Kinase Biology
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3552
Encompass
View Documents
LIFE TECHNOLOGIES
PV3552
25μg
Each for $1,120.00
Add to cart
 
Description

Description

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

For use in combination with Fluorescein-poly-GT (PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Specifications

Specifications

150kDa
Primary
Cell Analysis, Cell Viability, Drug Discovery and Development, Kinase Antibodies and Immunoassays, Pharma and Biopharma, Proliferation and Function, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation, Validation Kinase Biology
384-well plate
Signal transduction
1 Kit(s)
–20°C
FITC (Fluorescein), Tb (Terbium), lanthanide chelates
RUO
Primary
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit