Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP (TGF-beta 1) Antibody (7F6) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$206.50 - $499.50

Specifications

Antigen LAP (TGF-beta 1)
Clone 7F6
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry reported in scientific literature (PMID 30147399), Immunohistochemistry-Paraffin reported by customer review
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP222114SS
SDP
View Documents
Novus Biologicals
NBP222114SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP222114
SDP
View Documents
Novus Biologicals
NBP222114
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

LAP (TGF-beta 1)
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology
PBS with 0.02% Sodium Azide
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
TGFB1
IgG1
Ammonium sulfate precipitation
44.3 kDa
7F6
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry reported in scientific literature (PMID 30147399), Immunohistochemistry-Paraffin reported by customer review
Monoclonal
Precipitation
RUO
Human, Mouse, Rat
P01137
7040
Purified recombinant fragment of human TGF beta 1 (amino acids 62-197) expressed in E. coli. [UniProt# P01137]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.