Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP2 beta/TMPO Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568824
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8824 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8824 Supplier Bethyl Laboratories, Inc Supplier No. A304840AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

Lamina-associated polypeptide 2 (LAP2), isoform beta is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.

Specifications

Antigen LAP2 beta/TMPO
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TMPO
Gene Accession No. P42166
Gene Alias CMD1T, lamina-associated polypeptide 2, LAP2, LEM domain containing 4, LEMD4, PRO0868, thymopoietin beta, Thymopoietin isoform alpha, thymopoietin-related peptide isoform alpha, thymopoietin-related peptide isoforms beta/gamma, TP, TP alpha, TP beta/gamma, TPRP isoform alpha
Gene Symbols TMPO
Host Species Rabbit
Immunogen Between 225 and 275
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7112
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.