Learn More
LAP2 beta/TMPO Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | LAP2 beta/TMPO |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8827
![]() |
Bethyl Laboratories, Inc
A304841AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Lamina-associated polypeptide 2 (LAP2), isoform beta is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.Specifications
LAP2 beta/TMPO | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CMD1T, lamina-associated polypeptide 2, LAP2, LEM domain containing 4, LEMD4, PRO0868, thymopoietin beta, Thymopoietin isoform alpha, thymopoietin-related peptide isoform alpha, thymopoietin-related peptide isoforms beta/gamma, TP, TP alpha, TP beta/gamma, TPRP isoform alpha | |
TMPO | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P42166 | |
7112 | |
Between 300 and 350 | |
Primary | |
4° C | |
TMPO |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.