Learn More
Invitrogen™ LAP2 Monoclonal Antibody (RL29)

Description
MA1-075 has been successfully used in western blot and immunofluorescence applications to detect LAP2 from human and mouse samples. In western blot, this antibody predominantly detects the LAP2 delta subunit. The MA1-075 immunogen is full length rat LAP2, and has predicted reactivity with rat samples.
Specifications
Specifications
| Antigen | LAP2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | RL29 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Gene | TMPO |
| Gene Accession No. | P42166, Q61029, Q61033, Q62733 |
| Gene Alias | 5630400D24Rik; AI195756; AI606875; AW214352; AW547477; CMD1T; lamina-associated polypeptide 2; Lamina-associated polypeptide 2, isoform alpha; Lamina-associated polypeptide 2, isoform beta; Lamina-associated polypeptide 2, isoforms alpha/zeta; Lamina-associated polypeptide 2, isoforms beta/delta/epsilon/gamma; Lamina-associated polypeptide 2, isoforms beta/gamma; Lap2; LEM domain containing 4; LEMD4; PRO0868; Splenin; Thymopentin; Thymopoietin; Thymopoietin (lamina associated polypeptide 2); thymopoietin beta; Thymopoietin isoform alpha; Thymopoietin isoform beta; Thymopoietin isoforms alpha/zeta; Thymopoietin isoforms beta/delta/epsilon/gamma; Thymopoietin, isoforms beta/gamma; Thymopoietin-related peptide isoform alpha; thymopoietin-related peptide isoforms beta/gamma; TMPO; TP; TP alpha; TP alpha/zeta; TP beta; TP beta/delta/epsilon/gamma; TP beta/gamma; TP5; TPRP isoform alpha; TPRP isoforms beta/gamma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.