Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596154 Supplier Invitrogen™ Supplier No. PA596154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPEFLEDPSV LTKDKLKSEL VANNVTLPAG EQRKDVYVQL YLQHLTARNR PPLPAGTNSK GPPDFSSDEE REPTPVLGSG AAAAGRSRAA VGRKATKKTD KPRQEDKDDL DVTELTNEDL LDQLVKYGVN PGPIVGTTRK LYEKKLLKLR EQGTESRSST PLPTISSSAE NTRQNGSNDS DRYSDNEEDS KIELKLEKRE PLKGRAKTPV TLKQRRVEHN QSYSQAGITE TEWTSGSSKG GPLQALTRES TRGSRRTPRK RVETSEHFRI DGPVISESTP IAETIMASSN ESLVVNRVTG NFKHASPILP ITEFSDIPRR APKKPLTRAE VGEKTEERRV ERDILKEMFP YEASTPTGIS ASCRRPIKGA AGRPLELSDF RMEESFSSKY VPKYVPLADV KSEKTKKGRS; Positive Samples: 22Rv1, K-562, HepG2, 293T, SH-SY5Y, H460; Cellular Location: Chromosome, Nucleus.

In eukaryotic cells, the nuclear envelope consists of inner and outer membranes which define the border between the nucleus and cytoplasm. The inner and outer nuclear membranes are joined by large, supramolecular assemblies called nuclear pore complexes through which various molecules pass into and out of the nucleus. The inner membrane is lined by a filamentous meshwork called the nuclear lamina made up of proteins called lamins. The attachment of the lamins to the inner nuclear membrane appears to occur through their association with 4 related integral membrane proteins called Lamin Associated Polypeptides (LAPs). LAP 1A, 1B and 1C share a high degree of sequence homology while LAP 2 is distinct from the others. LAP 1A/B bind lamins A, C and B1, while LAP 2 binds lamin B1 only. In addition to lamin binding, LAP 2 has been shown to bind to chromosomes. Phosphorylation of LAP 2 during mitosis inhibits binding to both lamins and chromosomes suggesting that LAP 2 plays an important role in nuclear envelope re-assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TMPO
Gene Accession No. P42166, Q61029, Q61033, Q62733
Gene Alias 5630400D24Rik; AI195756; AI606875; AW214352; AW547477; CMD1T; lamina-associated polypeptide 2; Lamina-associated polypeptide 2, isoform alpha; Lamina-associated polypeptide 2, isoform beta; Lamina-associated polypeptide 2, isoforms alpha/zeta; Lamina-associated polypeptide 2, isoforms beta/delta/epsilon/gamma; Lamina-associated polypeptide 2, isoforms beta/gamma; Lap2; LEM domain containing 4; LEMD4; PRO0868; Splenin; Thymopentin; Thymopoietin; Thymopoietin (lamina associated polypeptide 2); thymopoietin beta; Thymopoietin isoform alpha; Thymopoietin isoform beta; Thymopoietin isoforms alpha/zeta; Thymopoietin isoforms beta/delta/epsilon/gamma; Thymopoietin, isoforms beta/gamma; Thymopoietin-related peptide isoform alpha; thymopoietin-related peptide isoforms beta/gamma; TMPO; TP; TP alpha; TP alpha/zeta; TP beta; TP beta/delta/epsilon/gamma; TP beta/gamma; TP5; TPRP isoform alpha; TPRP isoforms beta/gamma
Gene Symbols TMPO
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21917, 25359, 7112
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.