Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115494 Supplier Invitrogen™ Supplier No. PA5115494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LAP2 alpha.

In eukaryotic cells, the nuclear envelope consists of inner and outer membranes which define the border between the nucleus and cytoplasm. The inner and outer nuclear membranes are joined by large, supramolecular assemblies called nuclear pore complexes through which various molecules pass into and out of the nucleus. The inner membrane is lined by a filamentous meshwork called the nuclear lamina made up of proteins called lamins. The attachment of the lamins to the inner nuclear membrane appears to occur through their association with 4 related integral membrane proteins called Lamin Associated Polypeptides (LAPs). LAP 1A, 1B and 1C share a high degree of sequence homology while LAP 2 is distinct from the others. LAP 1A/B bind lamins A, C and B1, while LAP 2 binds lamin B1 only. In addition to lamin binding, LAP 2 has been shown to bind to chromosomes. Phosphorylation of LAP 2 during mitosis inhibits binding to both lamins and chromosomes suggesting that LAP 2 plays an important role in nuclear envelope re-assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TMPO
Gene Accession No. P42166, Q61029, Q61033, Q62733
Gene Alias 5630400D24Rik; AI195756; AI606875; AW214352; AW547477; CMD1T; lamina-associated polypeptide 2; Lamina-associated polypeptide 2, isoform alpha; Lamina-associated polypeptide 2, isoform beta; Lamina-associated polypeptide 2, isoforms alpha/zeta; Lamina-associated polypeptide 2, isoforms beta/delta/epsilon/gamma; Lamina-associated polypeptide 2, isoforms beta/gamma; Lap2; LEM domain containing 4; LEMD4; PRO0868; Splenin; Thymopentin; Thymopoietin; Thymopoietin (lamina associated polypeptide 2); thymopoietin beta; Thymopoietin isoform alpha; Thymopoietin isoform beta; Thymopoietin isoforms alpha/zeta; Thymopoietin isoforms beta/delta/epsilon/gamma; Thymopoietin, isoforms beta/gamma; Thymopoietin-related peptide isoform alpha; thymopoietin-related peptide isoforms beta/gamma; TMPO; TP; TP alpha; TP alpha/zeta; TP beta; TP beta/delta/epsilon/gamma; TP beta/gamma; TP5; TPRP isoform alpha; TPRP isoforms beta/gamma
Gene Symbols TMPO
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMPO(Accession P42166), corresponding to amino acid residues P402-P452.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21917, 25359, 7112
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.