Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lap2alpha, Mouse anti-Mouse, Clone: 8C10-1H11, Millipore Sigma™

Catalog No. MABF199725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF199725 25 μg
MABF1997100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF199725 Supplier MilliporeSigma Supplier No. MABF199725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Lap2alpha, clone 8C10-1H11, Cat. No. MABF1997, is a mouse monoclonal antibody that detects Lamina-associated polypeptide 2, isoforms alpha, and has been tested for use in Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), and Western Blotting.

Lamina-associated polypeptide 2, isoforms alpha (UniProt: Q61033; also known as Thymopoietin isoform alpha, TP alpha) is encoded by the Tmpo (also known as Lap2) gene (Gene ID: 21917) in murine species. LAP2alpha is a non-membrane bound, oligomeric nucleoplasmic protein that associates directly or indirectly with lamins at specific cell-cycle stages. It is expressed diffusely throughout the nucleus and is involved in the structural organization of the nucleus and in the post-mitotic nuclear assembly. It is shown to interact with A-type lamins to control gene expression, transcription, and chromatin organization. The N-terminal part of LAP2alpha contains two structurally independent, non-interacting domains: LEM (LAPEmerin-MAN1)-like domain that binds DNA and a LEM domain that interacts with BANF1. Its C-terminal domain forms a four-stranded coiled coil. Lamin A/C and LAP2alpha complexes predominantly co-occur in euchromatic genomic regions and are shown to be highly mobile. They can bind chromatin in a dynamic manner and are involved in establishing a chromatin environment that is permissive for epigenetic regulation. Loss of LAP2alpha has been linked to increased proliferation of tissue progenitor cells in epidermis, colon, and the hematopoietic system and delayed skeletal muscle differentiation. (Res.: Gesson, K et al., (2016). Genome Res. 26(4);462-473).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lap2alpha
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8C10-1H11
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Lap2alpha in Western Blotting applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Lap2alpha in Chromatin Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).Immunoprecipitation Analysis: A representative lot immunoprecipitated Lap2alpha in Immunoprecipitation applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).Immunofluorescence Analysis: A representative lot detected Lap2alpha in Immunofluorescence applications (Gesson, K., et. al. (2016). Genome Res. 26(4):462-73).
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Lamina-associated polypeptide 2 isoform alpha;Thymopoietin isoforms alpha;TP alpha
Gene Symbols Tmpo;Lap2
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 507 amino acids from the C-terminal region.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_035735
Test Specificity Clone 8C10-1H11 specifically detects Lamina-associated polypeptide 2 alpha (LAP2alpha) in murine species. It targets an epitope in the internal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.