Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556206 Supplier Invitrogen™ Supplier No. PA556206
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGRVVVRRLA VRRFGSRSLS TADMTKGLVL GIYSKEKEDD VPQFTSAGEN FDKLLAGKLR ETLNISGPPL KAGKTRTFYG LHQDFPSVVL VG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.

Leucine Aminopeptidase 3 (LAP3) is involved in the processing and regular turnover of intracellular proteins. It catalyzes the removal of unsubstituted N-terminal amino acids from various peptides. Release of an N-terminal amino acid, Xaa-/-Yaa-, in which Xaa is preferably Leu, but may be other amino acids including Pro although not Arg or Lys, and Yaa may be Pro. Amino acid amides and methyl esters are also readily hydrolyzed, but reated on arylamides are exceedingly low.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAP3
Gene Accession No. P28838
Gene Alias 2410015L10Rik; AA410100; Cytosol aminopeptidase; epididymis secretory protein Li 106; HEL-S-106; Lap; Lap3; LAP-3; LAPEP; leucine aminopeptidase 3; Leucyl aminopeptidase; Pep7; Pep-7; Peps; Pep-S; peptidase 7; peptidase S; Proline aminopeptidase; prolyl aminopeptidase
Gene Symbols LAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAP3. Recombinant protein control fragment (Product #RP-95809).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.