Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 146121AP150UL
Description
LAP3 is presumably involved in the processing and regular turnover of intracellular proteins. It catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.Release of an N-terminal amino acid, Xaa-/-Yaa-, in which Xaa is preferably Leu, but may be other amino acids including Pro although not Arg or Lys, and Yaa may be Pro.Specifications
| LAP3 | |
| Polyclonal | |
| Unconjugated | |
| LAP3 | |
| Cytosol aminopeptidase, LAP, LAP 3, LAP3, LAPEP, leucine aminopeptidase 3, Leucyl aminopeptidase, PEPS, Peptidase S, Proline aminopeptidase, Prolyl aminopeptidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289668, 51056, 66988 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P28838, Q68FS4, Q9CPY7 | |
| Lap3 | |
| LAP3 Fusion Protein Ag6205 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction