Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987928
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7928 Supplier Bioss Supplier No. BS7687R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Leucine Aminopeptidase 3 (LAP3) is involved in the processing and regular turnover of intracellular proteins. It catalyzes the removal of unsubstituted N-terminal amino acids from various peptides. Release of an N-terminal amino acid, Xaa-/-Yaa-, in which Xaa is preferably Leu, but may be other amino acids including Pro although not Arg or Lys, and Yaa may be Pro. Amino acid amides and methyl esters are also readily hydrolyzed, but reated on arylamides are exceedingly low.

Specifications

Antigen LAP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LAP3
Gene Accession No. P28838
Gene Alias 2410015L10Rik; AA410100; Cytosol aminopeptidase; epididymis secretory protein Li 106; HEL-S-106; Lap; LAP3; LAP-3; LAPEP; Leucine aminopeptidase 3; Leucyl aminopeptidase; Pep7; Pep-7; PEPS; Pep-S; peptidase 7; Peptidase S; Proline aminopeptidase; Prolyl aminopeptidase
Gene Symbols LAP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LAP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.