Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAPTM4A Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299293F
Description
LAPTM4A Polyclonal antibody specifically detects LAPTM4A in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
LAPTM4A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Golgi 4-transmembrane-spanning transporter MTP, HUMORF13, KIAA0108MBNT, LAPTM4lysosomal-associated transmembrane protein 4A, lysosomal protein transmembrane 4 alpha, lysosomal-associated protein transmembrane 4 alpha, membrane nucleoside transporter, Mtrp | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human LAPTM4A. | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
9741 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction