Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAPTM4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556731 Supplier Invitrogen™ Supplier No. PA556731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVTHPNSMPA VNIQYEVIGN YYSSERMADN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

May function in the transport of nucleosides and/or nucleoside derivatives between the cytosol and the lumen of an intracellular membrane-bound compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAPTM4A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAPTM4A
Gene Accession No. Q15012
Gene Alias AA286466; golgi 4-transmembrane spanning transporter; golgi 4-transmembrane-spanning transporter; golgi 4-transmembrane-spanning transporter MTP; HUMORF13; KIAA0108; LAPTM4; LAPTM4A; lysosomal protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4 alpha; lysosomal-associated protein transmembrane 4A; lysosomal-associated transmembrane protein 4A; lysosomal-associated transmembrane protein 4-like protein; MBNT; membrane nucleoside transporter; membrane transporter protein; mKIAA0108; MTP; Mtrp; similar to lysosomal-associated protein transmembrane 4 alpha; transmembrane glycoprotein; transporter protein; UNQ1846/PRO3574; wu:fb57c07; wu:fj99e08
Gene Symbols LAPTM4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAPTM4A. Recombinant protein control fragment (Product #RP-95971).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.