Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAPTM4B Antibody (CL5287), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | LAPTM4B |
---|---|
Clone | CL5287 |
Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
LAPTM4B Monoclonal specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LAPTM4B | |
Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
Mouse | |
Human | |
55353 | |
This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:DTTVLLPPYDDATVNGAAKE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL5287 | |
Monoclonal | |
Purified | |
Stem Cell Markers | |
LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta | |
LAPTM4B | |
IgG2b | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title