Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAPTM4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567016 Supplier Invitrogen™ Supplier No. PA567016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNSIQEYIRQL PPNFPYRDDV MSVNP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

LAPTM4B is a multi-pass membrane protein. It belongs to the LAPTM4/LAPTM5 transporter family. LAPTM4b has active role in disease progression of malignant cells and is involved in cell proliferation and multidrug resistance. The genetic polymorphism of LAPTM4B is a potential risk factor for the development of colon cancer and gastric cancer. The research results also indicated that LAPTM4B may be a clinically useful prognostic indicator for ovarian carcinoma and may play a role in human hepatocellular carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAPTM4B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAPTM4B
Gene Accession No. Q86VI4
Gene Alias C330023P13Rik; hypothetical protein 5; Laptm4b; LAPTM4beta; LC27; LOW QUALITY PROTEIN: lysosomal-associated transmembrane protein 4B; lysosomal associated protein transmembrane 4 beta; lysosomal protein transmembrane 4 beta; lysosomal-associated protein transmembrane 4B; lysosomal-associated transmembrane protein 4; lysosomal-associated transmembrane protein 4B; Lysosomal-associated transmembrane protein 4B-like protein; lysosome-associated transmembrane protein 4-beta; PSEC0001
Gene Symbols LAPTM4B
Host Species Rabbit
Immunogen Recombinant Human LAPTM4B. Recombinant protein control fragment (Product #RP-107657).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55353
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.