Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LAPTM4B Recombinant Protein

Catalog No. 89968720
Click to view available options
:
2 μg

Used for ELISA, Western Blotting (Recombinant Protein), Antibody Production and Protein Array

Sequence: MKMVAPWTRFYSNSCCLCCHVRTGTILLGVWYLIINAVVLLILLSALADPDQYNFSSSELGGVFEFMDDANMCIAIAISLLMILICAMATYGAYKQRAAWIIPFFCYQIFDFALNMLVAITVLIYPNSIQEYTRQLPPNFPYRDDVMSVNPTCLVLIILLFISIILTFKGYLISCVWNCYRYINGRNSSDVLVYVTSNDTTVLLPPYDDATVNGAAKEPPPPYVSA

Specifications

Accession Number AAH14129
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris HCl, 10 mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 55353
Molecular Weight (g/mol) 51 kDa
Name LAPTM4B (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Immunogen MKMVAPWTRFYSNSCCLCCHVRTGTILLGVWYLIINAVVLLILLSALADPDQYNFSSSELGGVFEFMDDANMCIAIAISLLMILICAMATYGAYKQRAAWIIPFFCYQIFDFALNMLVAITVLIYPNSIQEYTRQLPPNFPYRDDVMSVNPTCLVLIILLFISIILTFKGYLISCVWNCYRYINGRNSSDVLVYVTSNDTTVLLPPYDDATVNGAAKEPPPPYVSA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias LAPTM4beta, LC27
Common Name LAPTM4B
Gene Symbol LAPTM4B
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.