Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAPTM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562370 Supplier Invitrogen™ Supplier No. PA562370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKCMNSVEEK RNSKMLQKVV LPSYEEALSL PSKTP Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 66%.

This gene encodes a transmembrane receptor that is associated with lysosomes. The encoded protein, also known as E3 protein, may play a role in hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAPTM5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAPTM5
Gene Accession No. Q13571
Gene Alias CD40-ligand-activated specific transcripts; CLAST6; gcd-10; granule cell death-10; human retinoic acid-inducible E3 protein; KIAA0085; LAPTM5; lysosomal associated multispanning membrane protein 5; lysosomal multispanning membrane protein 5; lysosomal protein transmembrane 5; lysosomal-associated multitransmembrane protein 5; lysosomal-associated protein transmembrane 5; lysosomal-associated transmembrane protein 5; Retinoic acid-inducible E3 protein
Gene Symbols LAPTM5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAPTM5. Recombinant protein control fragment (Product #RP-101410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.