Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAPTM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571402
Description
LAPTM5 Polyclonal Antibody for Western Blot
This gene encodes a transmembrane receptor that is associated with lysosomes. The encoded protein, also known as E3 protein, may play a role in hematopoiesis.
Specifications
LAPTM5 | |
Polyclonal | |
Unconjugated | |
LAPTM5 | |
CD40-ligand-activated specific transcripts; CLAST6; gcd-10; granule cell death-10; human retinoic acid-inducible E3 protein; KIAA0085; LAPTM5; lysosomal associated multispanning membrane protein 5; lysosomal multispanning membrane protein 5; lysosomal protein transmembrane 5; lysosomal-associated multitransmembrane protein 5; lysosomal-associated protein transmembrane 5; lysosomal-associated transmembrane protein 5; Retinoic acid-inducible E3 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
89783 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
LAPTM5 | |
synthetic peptide directed towards the middle region of rat LAPTM5. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction