Learn More
Invitrogen™ LAPTM5 Polyclonal Antibody

Description
This antibody is predicted to react with bovine based on sequence homology.
Specifications
Specifications
| Antigen | LAPTM5 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | LAPTM5 |
| Gene Accession No. | Q13571 |
| Gene Alias | CD40-ligand-activated specific transcripts; CLAST6; gcd-10; granule cell death-10; human retinoic acid-inducible E3 protein; KIAA0085; LAPTM5; lysosomal associated multispanning membrane protein 5; lysosomal multispanning membrane protein 5; lysosomal protein transmembrane 5; lysosomal-associated multitransmembrane protein 5; lysosomal-associated protein transmembrane 5; lysosomal-associated transmembrane protein 5; Retinoic acid-inducible E3 protein |
| Gene Symbols | LAPTM5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.