Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAR4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565339 Supplier Invitrogen™ Supplier No. PA565339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRKPSYAEICQ RTSKEPPSSP LQPQKEQKPN TVGCGKEEKK LAEPAERYRE PPALKSTPGA PRDQRRPAGG RPSPSAMGKR LSR Highest antigen sequence identity to the following orthologs - mouse 78%, rat 87%.

The LAR4B gene, more accurately referred to as LARP4B (La Ribonucleoprotein Domain Family Member 4B), is associated with RNA-binding proteins that are crucial for various aspects of RNA metabolism, including RNA stabilization, translation, and cellular stress responses. LARP4B belongs to the La-related protein family, which plays significant roles in maintaining RNA stability and enhancing translation efficiency of certain mRNAs. It has been identified as a target in cancer research due to its involvement in tumor biology, particularly regulating tumor suppressive pathways and interacting with microRNA networks. Studies have shown that LARP4B expression is downregulated in prostate cancer tissues, indicating its potential role as a tumor suppressor. Moreover, it is linked with liver cancer and survival rates, reflecting its importance in cellular growth and proliferation. The regulation of LARP4B and its interactions with other cellular components underline its potential therapeutic implications for cancer treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAR4B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LARP4B
Gene Accession No. Q92615
Gene Alias A630096F19; AI256361; D13Wsu64e; KIAA0217; La ribonucleoprotein 4B; La ribonucleoprotein domain family member 4B; La ribonucleoprotein domain family member 5; La ribonucleoprotein domain family, member 4B; La ribonucleoprotein domain family, member 5; la-related protein 4B; La-related protein 5; LARP4B; Larp5; RGD1310713
Gene Symbols LARP4B
Host Species Rabbit
Immunogen Recombinant Human LAR4B. Recombinant protein control fragment (Product #RP-104757).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.