Learn More
LARG Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301960AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli working through G protein-coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein has been observed to form a myeloid/lymphoid fusion partner in acute myeloid leukemia. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2014].Specifications
LARG | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NZN5 | |
ARHGEF12 | |
Between 1494 and 1544 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ARHGEF12 | |
KIAA0382, LARG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23365 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.