Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LARGE Antibody (LARGE-02), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262211PE
Description
LARGE Monoclonal specifically detects LARGE in Human samples. It is validated for Flow Cytometry.Specifications
LARGE | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
LARGE | |
Recombinant fragment of human LARGE1 (amino acids 35-142) | |
0.1 mL | |
9215 | |
Human | |
IgG2b |
Flow Cytometry | |
LARGE-02 | |
Flow Cytometry | |
Acetylglucosaminyltransferase-like 1A, EC 2.4, glycosyltransferase-like protein LARGE1, KIAA0609acetylglucosaminyltransferase-like protein, LARGE1, like-acetylglucosaminyltransferase, like-glycosyltransferase, MDC1D, MDDGA6, MDDGB6 | |
Mouse | |
Protein A purified | |
Primary | |
The antibody MEM-65 recognizes an unique epitope of CD2, a 50 kDa glycoprotein present on the human peripheral blood T-lymphocytes and NK cells; also expressed by all thymocytes. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction