Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Large Bore Male Luer with 5/16 in. ID - CrystalVu - QC
SDP

Catalog No. 01000183 Shop All Fisherbrand Products

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Large bore luers offer a higher flow rates than standard luers. 

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • RoHS Compliant
  • Gamma Stable
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Product Type Large Bore Luer
Description Class 7 Cleanroom Manufactured
Material CrystalVu™ (CVU)
For Use With (Application) Medical, Pharmaceutical, Industrial applications
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Transparent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.