Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARGE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519292
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519292 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519292 Supplier Invitrogen™ Supplier No. PA519292
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARGE
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Large1
Gene Accession No. O95461
Gene Alias Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; BPFD#36; enr; eosinophil granule major basic protein 1; fg; froggy; glycosyltransferase-like 1A; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; Gyltl1a; KIAA0609; large; LARGE xylosyl- and glucuronyltransferase 1; Large1; like-acetylglucosaminyltransferase; like-glycosyltransferase; MBP1; Mbp-1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd; myodystrophy; prepro major basic protein 1; Xylosyltransferase LARGE
Gene Symbols Large1
Host Species Goat
Immunogen Synthetic peptide sequence (SEADVNSENLQKQ) corresponding to the internal amino acids of LARGE (aa 421-434).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9215
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.