Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARGE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578393 Supplier Invitrogen™ Supplier No. PA578393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse liver Predicted Reactivity: Human (100%), Rhesus Monkey (100%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARGE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Large1
Gene Accession No. O95461, Q9Z1M7
Gene Alias Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Beta-1,3-glucuronyltransferase LARGE; BPFD#36; enr; eosinophil granule major basic protein 1; fg; froggy; glycosyltransferase-like 1A; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; glycosyltransferase-like protein LARGE1; LARGE xylosyl- and glucuronyltransferase 1; Gyltl1a; KIAA0609; large; LARGE xylosyl- and glucuronyltransferase 1; Large1; like-acetylglucosaminyltransferase; like-glycosyltransferase; MBP1; Mbp-1; MDC1D; MDDGA6; MDDGB6; mKIAA0609; myd; myodystrophy; prepro major basic protein 1; Xylosyltransferase LARGE
Gene Symbols Large1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LARGE. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16795, 361368, 9215
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.