Learn More
Mopec™ Large Slide Drying Hotplate Lid
SDP
Catalog No. 22445579
Click to view available options
Product Type:
Hotplate Lid
Large Slide Drying Hotplate is designed to quickly and efficiently dry paraffin tissue sections. The rapid heating system ensures the hotplate is ready to use in less time, while the digital temperature control allows for accurate control of temperatures.
  • For use with the Mopec™ Large Slide Drying Hotplate
  • Helps reduce drying time and improves temperature control
Specifications
Product Type Hotplate Lid
Material Stainless Steel
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.