Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LARP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535912
Description
Recommended positive controls: IMR32, SK-N-AS. Predicted reactivity: Mouse (98%), Xenopus laevis (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNA-binding protein that regulates the translation of specific target mRNA species downstream of the mTORC1 complex, in function of growth signals and nutrient availability (PubMed:20430826, PubMed:23711370, PubMed:24532714, PubMed:25940091, PubMed:28650797, PubMed:28673543, PubMed:29244122). Interacts on the one hand with the 3' poly-A tails that are present in all mRNA molecules, and on the other hand with the 7-methylguanosine cap structure of mRNAs containing a 5' terminal oligopyrimidine (5'TOP) motif, which is present in mRNAs encoding ribosomal proteins and several components of the translation machinery (PubMed:23711370, PubMed:25940091, PubMed:28650797, PubMed:29244122, PubMed:26206669, PubMed:28379136). The interaction with the 5' end of mRNAs containing a 5'TOP motif leads to translational repression by preventing the binding of EIF4G1 (PubMed:25940091, PubMed:28650797, PubMed:29244122, PubMed:28379136). When mTORC1 is activated, LARP1 is phosphorylated and dissociates from the 5' untranslated region (UTR) of mRNA (PubMed:25940091, PubMed:28650797). Does not prevent binding of EIF4G1 to mRNAs that lack a 5'TOP motif (PubMed:28379136). Interacts with the free 40S ribosome subunit and with ribosomes, both monosomes and polysomes (PubMed:20430826, PubMed:24532714, PubMed:25940091, PubMed:28673543). Under normal nutrient availability, interacts primarily with the 3' untranslated region (UTR) of mRNAs encoding ribosomal proteins and increases protein synthesis (PubMed:23711370, PubMed:28650797). Associates with actively translating ribosomes and stimulates translation of mRNAs containing a 5'TOP motif, thereby regulating protein synthesis, and as a consequence, cell growth and proliferation (PubMed:20430826, PubMed:24532714). Stabilizes mRNAs species with a 5'TOP motif, which is required to prevent apoptosis (PubMed:20430826, PubMed:23711370, PubMed:25940091, PubMed:28673543). [UniProt].
Specifications
LARP1 | |
Polyclonal | |
Unconjugated | |
Larp1 | |
0031/08; 0225/29; 1810024J12Rik; 3110040D16Rik; BcDNA:LD02611; CG14065; CG14066; CG42551; CG42551-PA; CG42551-PB; CG42551-PD; CG42551-PE; CG42551-PF; CG42551-PG; CG42551-PH; dlarp; Dmel\CG42551; Dmel_CG42551; drosophila La related protein; KIAA0731; l(3)S003108; l(3)S022529; La related protein; La ribonucleoprotein domain family member 1; La ribonucleoprotein domain family, member 1; La-related protein; la-related protein 1; Larp; Larp1; larp-PA; larp-PB; larp-PD; larp-PE; larp-PF; larp-PG; larp-PH; lethal (3) S003108; lethal (3) S022529; meteor; mKIAA0731; RGD1306683 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23367 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6PKG0 | |
Larp1 | |
Recombinant fragment corresponding to a region within amino acids 661 and 1019 of LARP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction