Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARP1 Rabbit anti-Drosophila, Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728948 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8948 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8948 Supplier Proteintech Group Inc Supplier No. 137081AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. Associates with the mRNA 5′cap in an MTOR-dependent manner and associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. Associates with actively translating ribosomes via interaction with PABPC1/PABP and stimulates translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. Positively regulates the replication of dengue virus (DENV).

Specifications

Antigen LARP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LARP1
Gene Accession No. Q6PKG0, Q6ZQ58
Gene Alias KIAA0731, La related protein 1, LARP, LARP1
Gene Symbols larp, Larp1
Host Species Rabbit
Immunogen LARP1 Fusion Protein Ag4645
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23367, 303158, 53567, 73158
Target Species Drosophila, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.