Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LARP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | LARP2 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
LARP2 Polyclonal specifically detects LARP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LARP2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55132 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KHKWVPLHLDVVRSESQERPGSRNSSRCQPEANKPTHNNRRNDTRSWKRDREKRDDQDDVSSVRSEGGNIRGSF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
DKFZp434K245, DKFZp686E0316, DKFZp686L13217, FLJ10378, La ribonucleoprotein domain family member 1B, La ribonucleoprotein domain family member 2, La ribonucleoprotein domain family, member 1B, La ribonucleoprotein domain family, member 2, la-related protein 1B, La-related protein 2, LARP2, MGC117277, MGC75174 | |
LARP1B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title