Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557593 Supplier Invitrogen™ Supplier No. PA557593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KHKWVPLHLD VVRSESQERP GSRNSSRCQP EANKPTHNNR RNDTRSWKRD REKRDDQDDV SSVRSEGGNI RGSF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

LARP2 encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LARP1B
Gene Accession No. Q659C4
Gene Alias 1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04
Gene Symbols LARP1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LARP2. Recombinant protein control fragment (Product #RP-96662).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55132
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.