Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518653
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518653 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518653 Supplier Invitrogen™ Supplier No. PA518653
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

LARP2 encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP2
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene LARP1B
Gene Accession No. Q659C4
Gene Alias 1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04
Gene Symbols LARP1B
Host Species Goat
Immunogen Synthetic peptide sequence (QGVLHIPKKDLTDE) corresponding to the internal amino acids of LARP2 (aa 553-567).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55132
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.