Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LARP4 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222254PE
Description
LARP4 Polyclonal antibody specifically detects LARP4 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
LARP4 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
c-Mpl binding protein, DKFZp686E039, La ribonucleoprotein domain family member 4, La ribonucleoprotein domain family, member 4, la-related protein 4, MGC74631, PP13296 | |
The immunogen this antibody was made to, maps to a region between residue 674 and 724 of human La Ribonucleoprotein Domain Family, Member 4 using the numbering given in entry NP_443111.4 (GeneID 113251). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
113251 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction