Learn More
Invitrogen™ LARP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113720
Description
Immunogen sequence: VSPTKNEDNG APENSVEKPH EKPEARASKD YSGFRGNIIP RGAAGKIREQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%.
LARP4 gene ontology annotations related to this gene include RNA binding.
Specifications
| LARP4 | |
| Polyclonal | |
| Unconjugated | |
| LARP4 | |
| c-Mpl binding protein; DXErtd793; DXErtd793e; La ribonucleoprotein domain family member 4; La ribonucleoprotein domain family, member 4; la-related protein 4; la-related protein 4-like; Larp4; PP13296 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113251 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q71RC2 | |
| LARP4 | |
| Synthetic peptide directed towards the middle region of human LARP4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.