Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541881 Supplier Invitrogen™ Supplier No. PA541881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MGTQEKSPGT SPLLSRKMQT ADGLPVGVLR LPRGPDNTRG FHGHERSRAC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 86%.

LARP6 is a member of the La ribonucleoprotein domain family, which all contain one characteristic La motif. LARP6 shuttles between the cytoplasm and nucleus. It is proposed that members of this family play a role in translation and mRNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP6
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LARP6
Gene Accession No. Q8BN59, Q9BRS8
Gene Alias 5430431G03Rik; acheron; Achn; AI552438; death-associated LA motif protein; La ribonucleoprotein 6, translational regulator; La ribonucleoprotein domain family member 6; La ribonucleoprotein domain family, member 6; la-related protein 6; Larp6; RGD1308414
Gene Symbols LARP6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LARP6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55323, 67557
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.