Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106330 Supplier Invitrogen™ Supplier No. PA5106330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LARP7.

LARP7 gene ontology annotations related to this gene include RNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LARP7
Gene Accession No. Q05CL8, Q4G0J3, Q5XI01
Gene Alias ALAZS; C330027G06Rik; D3Wsu161e; DKFZP564K112; HDCMA18P; La ribonucleoprotein 7, transcriptional regulator; La ribonucleoprotein domain family member 7; La ribonucleoprotein domain family, member 7; la-related protein 7; la-related protein 7; LOW QUALITY PROTEIN: la-related protein 7; Larp7; PIP7S; P-TEFb-interaction protein for 7SK stability; Unknown (protein for IMAGE:8171460); Unknown (protein for IMAGE:8281412)
Gene Symbols LARP7
Host Species Rabbit
Immunogen A synthesized peptide derived from human LARP7(Accession Q4G0J3), corresponding to amino acid residues L524-K574.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28036, 51574, 686883
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.