Learn More
LARP7 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303724A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function remains unknown.Specifications
LARP7 | |
Polyclonal | |
Unconjugated | |
LARP7 | |
ALAZS, DKFZP564K112, HDCMA18P, La ribonucleoprotein domain family member 7, La ribonucleoprotein domain family, member 7, PIP7S, P-TEFb-interaction protein for 7SK stability | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28036, 51574 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q05CL8, Q4G0J3 | |
LARP7 | |
Between 532 and 582 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.