Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595627
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595627 Supplier Invitrogen™ Supplier No. PA595627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HL-60 whole cell, human K562 whole cell, human THP-1 whole cell, human Caco-2 whole cell, rat liver tissue, mouse brain tissue, mouse liver tissue, mouse NIH3T3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene LARS1
Gene Accession No. Q8BMJ2, Q9P2J5
Gene Alias 2310045K21Rik; 3110009L02Rik; AW536573; cytoplasmic leucyl-tRNA synthetase; cytosolic leucyl-tRNA synthetase; hr025Cl; HSPC192; ILFS1; KIAA1352; Lars; LARS1; leucine translase; leucine tRNA ligase 1, cytoplasmic; leucine-tRNA ligase; leucine--tRNA ligase, cytoplasmic; Leucyl-tRNA synthetase; leucyl-tRNA synthetase, cytoplasmic; LEURS; LEUS; LFIS; LRS; mKIAA1352; PIG44; proliferation-inducing gene 44; RNTLS
Gene Symbols LARS1
Host Species Rabbit
Immunogen E.coli-derived human Leucyl tRNA synthetase/LARS recombinant protein (Q628-Q855).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107045, 291624, 51520
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.