Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522086

Catalog No. PIPA522086


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (86%), Rat (89%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LARS2
Polyclonal
Unconjugated
LARS2
AI035546; Kiaa0028; LARS2; leucine translase; leucine tRNA ligase 2, mitochondrial; leucine tRNA ligase 2, mitocondrial; leucine--tRNA ligase; Leucyl-tRNA synthetase; leucyl-tRNA synthetase 2; leucyl-tRNA synthetase 2, mitochondrial; leucyl-tRNA synthetase, mitochondrial; LEURS; mtLeuRS; PRLTS4; Probable leucine--tRNA ligase, mitochondrial; probable leucyl-tRNA synthetase, mitochondrial
Rabbit
Antigen Affinity Chromatography
RUO
23395
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q15031
LARS2
Recombinant fragment corresponding to a region within amino acids 181 and 458 of LARS2 (Uniprot ID#Q15031).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.