Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568646 Supplier Invitrogen™ Supplier No. PA568646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Horse: 85%; Human: 100%.

Involved in the biogenesis of the 60S ribosomal subunit. Required for maturation of the 28S rRNA. Functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex; the 5FMC complex is recruited to ZNF148 by methylated CHTOP, leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAS1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Las1l
Gene Accession No. Q9Y4W2
Gene Alias 1810030A06Rik; 5830482G23Rik; dJ475B7.2; LAS1 like ribosome biogenesis factor; LAS1 like, ribosome biogenesis factor; LAS1L; Las1-like; LAS1-like (S. cerevisiae); LAS1-like, ribosome biogenesis factor; MSTP060; Protein LAS1 homolog; RGD1565755; Ribosomal biogenesis protein LAS1L
Gene Symbols Las1l
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LAS1L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81887
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.