Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAS2 Antibody (OTI2B2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272452IR
Description
LAS2 Monoclonal specifically detects LAS2 in Human samples. It is validated for Western Blot.Specifications
LAS2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C18orf54 | |
Full length recombinant protein of human LAS2 (NP_775800) produced in E.coli. | |
0.1 mL | |
162681.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2B2 | |
Western Blot | |
C18orf54, chromosome 18 open reading frame 54, hypothetical protein LOC162681, LAS2, MGC33382 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction