Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
lasB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5144408

Description
Cleaves host elastin, collagen, IgG, and several complement components as well as endogenous pro-aminopeptidase (PubMed:11533066). Autocatalyses processing of its pro-peptide (PubMed:9642203, PubMed:1744034). Processes the pro-peptide of pro-chitin-binding protein (cbpD) (PubMed:9642203). Involved in the pathogenesis of P.aeruginosa infections.
Specifications
lasB | |
Polyclonal | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P14756 | |
lasB | |
Recombinant Pseudomonas aeruginosa (strain ATCC 15692/DSM 22644/CIP 104116/JCM 14847/LMG 12228/1C/PRS 101/PAO1) ELAS protein (198-498aa) | |
100 μL | |
Primary | |
Bacteria | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
lasB | |
Elastase, lasB PA3724 | |
Rabbit | |
Protein G | |
RUO | |
880368 | |
-20°C or -80°C if preferred | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction