Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LASP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219334
Description
LASP1 Polyclonal specifically detects LASP1 in Human samples. It is validated for Western Blot.Specifications
LASP1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
LASP-1, LIM and SH3 domain protein 1, LIM and SH3 protein 1, Metastatic lymph node gene 50 protein, MLN 50, MLN50Lasp-1 | |
Rabbit | |
30 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Rabbit (100%), Rat (100%), Rhesus Monkey (100%), Xenopus tropicalis (92%), Xenopus laevis (92%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q14847 | |
LASP1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human LASP1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
3927 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction