Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528711 Supplier Invitrogen™ Supplier No. PA528711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: LASP1 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Rabbit (100%), Chicken (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LASP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Lasp1
Gene Accession No. Q14847
Gene Alias AA408629; Def-4; Lasp1; LASP-1; lasp1 {ECO:0000312; LIM and SH3 domain protein 1; LIM and SH3 protein 1; metastatic lymph node gene 50 protein; MLN 50; MLN50; RGD:68408}; SH3P6; Tg(11Col1a1)NG; Tg(Col1a1-lacZ)1Ngma
Gene Symbols Lasp1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human LASP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.