Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579581
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579581 Supplier Invitrogen™ Supplier No. PA579581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Rat Spleen Tissue, Rat Intestine Tissue, JURKAT Cell, MCF-7 Cell, A431 Cell, HELA Cell, 293T Cell. IHC: Human Lung Cancer Tissue. ICC/IF: A549 Cell.

This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LASP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Lasp1
Gene Accession No. Q14847, Q61792, Q99MZ8
Gene Alias AA408629; Def-4; Lasp1; LASP-1; lasp1 {ECO:0000312; LIM and SH3 domain protein 1; LIM and SH3 protein 1; metastatic lymph node gene 50 protein; MLN 50; MLN50; RGD:68408}; SH3P6; Tg(11Col1a1)NG; Tg(Col1a1-lacZ)1Ngma
Gene Symbols Lasp1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human LASP1 (16-30aa EKVNCLDKFWHKACF).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16796, 29278, 3927
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.