Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lass1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA112918
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA112918 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA112918 Supplier Invitrogen™ Supplier No. PA112918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is hypothalamus lysate.

This gene encodes a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lass1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene CERS1
Gene Accession No. P27544
Gene Alias Ceramide synthase 1; CERS1; EPM8; LAG1; LAG1 homolog, ceramide synthase 1; LAG1 longevity assurance homolog 1; LASS1; longevity assurance (LAG1, S. cerevisiae) homolog 1; longevity assurance gene 1 protein homolog 1; longevity assurance homolog 1; longevity assurance-like protein 1; MGC90349; protein UOG-1; UOG1; upstream of GDF1
Gene Symbols CERS1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the human Lass1 protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10715
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.